CAS RN 947-04-6
CAS RN 947-04-6
IUPAC Nome:
1-azacyclotridecan-2-one
Sinonimi:
nylon-12 laurolactam
Peso molecolare (g/mol):
197.32
Formula molecolare:
C12H23NO
InChi Key:
JHWNWJKBPDFINM-UHFFFAOYSA-N
SMILES:
O=C1CCCCCCCCCCCN1
1
–
1
di
1
risultati
1
Lauryl lactam 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality