CAS RN 947-62-6
IUPAC Nome:
2-methoxynaphthalene-1-carboxylic acid
Sinonimi:
2-methoxynaphthalene-1-carboxylic acid
Peso molecolare (g/mol):
202.21
Formula molecolare:
C12H10O3
InChi Key:
OSTYZAHQVPMQHI-UHFFFAOYSA-N
SMILES:
COC1=CC=C2C=CC=CC2=C1C(O)=O