CAS RN 94840-08-1
CAS RN 94840-08-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
2,3-O-Isopropylidene-L-gulonolactone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.