CAS RN 948851-85-2
CAS RN 948851-85-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
(8Z,11Z,14Z,17Z)-Methyl 5,6-Dihydroxyicosa-8,11,14,17-tetraenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(8Z,11Z,14Z,17Z)-Methyl 5,6-Dihydroxyicosa-8,11,14,17-tetraenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(8Z,11Z,14Z,17Z)-Methyl 5,6-Dihydroxyicosa-8,11,14,17-tetraenoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.