CAS RN 950762-95-5
CAS RN 950762-95-5
IUPAC Nome:
N-hydroxy-1-[(4-methoxyphenyl)methyl]-1H-indole-6-carboxamide
Sinonimi:
N-hydroxy-1-[(4-methoxyphenyl)methyl]indole-6-carboxamide
Peso molecolare (g/mol):
296.33
Formula molecolare:
C17H16N2O3
InChi Key:
AJRGHIGYPXNABY-UHFFFAOYSA-N
SMILES:
COC1=CC=C(CN2C=CC3=CC=C(C=C23)C(=O)NO)C=C1
1
–
2
di
2
risultati
1
2
PCI-34051, MedChemExpress
MedChemExpress PCI-34051 is a potent and selective HDAC8 inhibitor with IC50 of 10 nM, with >200-fold selectivity over the other HDAC isoforms.