CAS RN 951650-22-9
CAS RN 951650-22-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
NS11394, MedChemExpress
MedChemExpress NS11394 is an orally active and unique subtype-selective GABAA positive allosteric receptor (PAM), with a Ki of ∼0.5 nM. NS11394 shows a selectivity profile in the order of GABAA-5 > α3 > α2 > α1-containing receptors. NS11394 has anxiolytic and anti-inflammatory properties.