CAS RN 95522-45-5
CAS RN 95522-45-5
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Colestilan, TRC
CAS: 95522-45-5 Formula molecolare: C4H6N2 . C3H5ClO Molecular Weight (g/mol): 174.63 Sinonimo: 2-Methyl-1H-imidazole polymer with 2-(Chloromethyl)oxirane,(Chloromethyl)oxirane polymer with 2-methyl-1H-imidazole,Cholebine,Colestilan Chloride,Colestimide,Epichlorohydrin-2-methylimidazole Copolymer,MCI 196 IUPAC Name: 2-(chloromethyl)oxirane 2-methyl-1H-imidazole SMILES: CC1=NC=CN1.ClCC2CO2