CAS RN 95617-09-7
CAS RN 95617-09-7
IUPAC Nome:
2-{4-[(6-chloro-1,3-benzoxazol-2-yl)oxy]phenoxy}propanoic acid
Sinonimi:
fenoxaprop fenoxaprop-P
Peso molecolare (g/mol):
333.72
Formula molecolare:
C16H12ClNO5
InChi Key:
MPPOHAUSNPTFAJ-UHFFFAOYNA-N
SMILES:
CC(OC1=CC=C(OC2=NC3=CC=C(Cl)C=C3O2)C=C1)C(O)=O
1
–
3
di
3
risultati
1
Fenoxaprop, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Fenoxaprop, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Fenoxaprop, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.