CAS RN 959396-07-7
CAS RN 959396-07-7
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
2-(1,1-Dimethylethyl)-3-hydroxy-17-oxoestra-1,3,5(10)-triene-4-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-(1,1-Dimethylethyl)-3-hydroxy-17-oxoestra-1,3,5(10)-triene-4-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-(1,1-Dimethylethyl)-3-hydroxy-17-oxoestra-1,3,5(10)-triene-4-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.