CAS RN 96-04-8
IUPAC Nome:
heptane-2,3-dione
Sinonimi:
heptane-2,3-dione
Peso molecolare (g/mol):
128.17
Formula molecolare:
C7H12O2
InChi Key:
FJPGAMCQJNLTJC-UHFFFAOYSA-N
SMILES:
CCCCC(=O)C(C)=O
1
–
1
di
1
risultati
1
2,3-Heptanedione, 97+%
CAS: 96-04-8 Formula molecolare: C7H12O2 Molecular Weight (g/mol): 128.171 Numero MDL: MFCD00036550 InChI Key: FJPGAMCQJNLTJC-UHFFFAOYSA-N Sinonimo: 2,3-heptanedione,acetyl valeryl,valerylacetyl,acetylvaleryl,acetyl pentanoyl,unii-dk55dde86p,dk55dde86p,benzil-related compound, 47,heptanedione,fema no. 2543 PubChem CID: 60983 IUPAC Name: heptane-2,3-dione SMILES: CCCCC(=O)C(=O)C