CAS RN 96201-88-6
CAS RN 96201-88-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
Tocris Bioscience™ Brequinar sodium
Potent and selective DHODH inhibitor.
2
Brequinar Sodium, TRC
CAS: 96201-88-6 Formula molecolare: C23H14F2NO2Na Molecular Weight (g/mol): 397.35 Sinonimo: 6-Fluoro-2-(2'-fluoro[1,1'-biphenyl]-4-yl)-3-methyl-4-quinolinecarboxylic Acid Sodium,Biphenquinate Sodium,DuP 785,NSC 368390 IUPAC Name: sodium;6-fluoro-2-[4-(2-fluorophenyl)phenyl]-3-methylquinoline-4-carboxylate SMILES: BrCC1=C(C2=CC=CC=C2)C=CC=C1