CAS RN 96315-08-1
CAS RN 96315-08-1
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
1-(5-Methyl-2-nitrophenoxy)-2-(2-nitrophenoxy)ethane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-(5-Methyl-2-nitrophenoxy)-2-(2-nitrophenoxy)ethane, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.