CAS RN 96861-65-3
CAS RN 96861-65-3
IUPAC Nome:
3-amino-6-chloro-N-(diaminomethylidene)-5-[methyl(2-methylpropyl)amino]pyrazine-2-carboxamide
Sinonimi:
3-amino-6-chloro-N-(diaminomethylidene)-5-[methyl(2-methylpropyl)amino]pyrazine-2-carboxamide
Peso molecolare (g/mol):
299.76
Formula molecolare:
C11H18ClN7O
InChi Key:
RVIUMPLAOXSSGN-UHFFFAOYSA-N
SMILES:
CC(C)CN(C)C1=NC(N)=C(N=C1Cl)C(=O)N=C(N)N
1
–
3
di
3
risultati
1
5-(N-Methyl-N-isobutyl)-Amiloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-(N-Methyl-N-isobutyl)-Amiloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
5-(N-Methyl-N-isobutyl)-Amiloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.