CAS RN 96946-44-0
CAS RN 96946-44-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
(1R,2R,1’R,2’S)-Atracurium Besylate, >90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1R,2R,1’R,2’S)-Atracurium Besylate, >90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(1R,2R,1’R,2’S)-Atracurium Besylate, >90%, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.