CAS RN 97732-82-6
CAS RN 97732-82-6
IUPAC Nome:
(propan-2-yl)benzene
Sinonimi:
cumene isopropilbenzene cumene (substance) 2-fenilpropano rcra waste number U055 cumeen 2-fenyl-propaan isopropylbenzeen
Peso molecolare (g/mol):
120.2
Formula molecolare:
C9H12
InChi Key:
RWGFKTVRMDUZSP-UHFFFAOYSA-N
SMILES:
CC(C)C1=CC=CC=C1
1
–
1
di
1
risultati
1
2-Phenylpropane-d12, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.