CAS RN 98-66-8
CAS RN 98-66-8
IUPAC Nome:
4-chlorobenzene-1-sulfonic acid
Sinonimi:
4-chlorobenzenesulfonic acid
Peso molecolare (g/mol):
192.61
Formula molecolare:
C6H5ClO3S
InChi Key:
RJWBTWIBUIGANW-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)C1=CC=C(Cl)C=C1
1
–
3
di
3
risultati
1
4-Chlorobenzenesulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Chlorobenzenesulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Chlorobenzenesulfonic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.