CAS RN 98014-14-3
CAS RN 98014-14-3
IUPAC Nome:
bis(2,2'-bipyridine) dichlororuthenium hydrate
Sinonimi:
bis(bipyridyl) dichlororuthenium hydrate
Peso molecolare (g/mol):
502.36
Formula molecolare:
C20H18Cl2N4ORu
InChi Key:
XBOQABVNRXVAKI-UHFFFAOYSA-L
SMILES:
O.Cl[Ru]Cl.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1
1
–
1
di
1
risultati
1
Promozioni disponibili
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%
CAS: 98014-14-3 Formula molecolare: C20H16Cl2N4Ru Molecular Weight (g/mol): 484.35 Numero MDL: MFCD00150005 InChI Key: MGAJEYXQYJBLQL-UHFFFAOYSA-L Sinonimo: cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate,bis bipyridyl dichlororuthenium hydrate,2,2'-bipyridine-dichlororuthenium-water 2/1/1,cis-dichlorobis 2,2'-bipyridine ruthenium ii,cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate,cis-bis 2,2-bipyridine dichlororuthenium ii hydrate,cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate,cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate PubChem CID: 16211844 SMILES: Cl[Ru]Cl.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1