CAS RN 98027-27-1
IUPAC Nome:
5-bromothiophene-2-carbohydrazide
Sinonimi:
5-bromothiophene-2-carbohydrazide
Peso molecolare (g/mol):
221.07
Formula molecolare:
C5H5BrN2OS
InChi Key:
OFXZACLPQYOIAV-UHFFFAOYSA-N
SMILES:
NNC(=O)C1=CC=C(Br)S1
1
–
1
di
1
risultati
1
5-Bromothiophene-2-carboxylic acid hydrazide, 97%
CAS: 98027-27-1 Formula molecolare: C5H5BrN2OS Molecular Weight (g/mol): 221.07 Numero MDL: MFCD01310798 InChI Key: OFXZACLPQYOIAV-UHFFFAOYSA-N Sinonimo: 5-bromo-2-thiophenecarboxylic acid hydrazide,5-bromo-thiophene-2-carboxylic acid hydrazide,5-bromo-thiophene-2-carboxylic acidhydrazide,5-bromothiophene-2-carboxylic acid hydrazide,2-thiophenecarboxylic acid, 5-bromo-, hydrazide,pubchem16117,ksc488m6p,5-bromo-2-thiophenecarboxylic hydrazide,2-bromothiophene-5-carbohydrazide,2-thiophenecarboxylicacid, 5-bromo-, hydrazide PubChem CID: 2063863 IUPAC Name: 5-bromothiophene-2-carbohydrazide SMILES: NNC(=O)C1=CC=C(Br)S1