CAS RN 98786-86-8
CAS RN 98786-86-8
IUPAC Nome:
3-amino-5-bromo-1,2-dihydropyridin-2-one
Sinonimi:
3-amino-5-bromo-1H-pyridin-2-one
Peso molecolare (g/mol):
189.01
Formula molecolare:
C5H5BrN2O
InChi Key:
ULOVLVWNSQNABA-UHFFFAOYSA-N
SMILES:
NC1=CC(Br)=CNC1=O
1
–
1
di
1
risultati
1
3-Amino-5-bromo-2-hydroxypyridine, 95%, Thermo Scientific Chemicals
CAS: 98786-86-8 Formula molecolare: C5H5BrN2O Molecular Weight (g/mol): 189.01 Numero MDL: MFCD03840438 InChI Key: ULOVLVWNSQNABA-UHFFFAOYSA-N Sinonimo: 3-amino-5-bromopyridin-2-ol,3-amino-5-bromo-2-hydroxypyridine,2-hydroxy-3-amino-5-bromopyridine,3-amino-5-bromopyridin-2 1h-one,3-amino-5-bromo-2-pyridinol,3-amino-5-bromo-pyridin-2-ol,2 1h-pyridinone, 3-amino-5-bromo,pubchem6513,pubchem18742,acmc-20a0ks PubChem CID: 13434515 IUPAC Name: 3-amino-5-bromo-1H-pyridin-2-one SMILES: NC1=CC(Br)=CNC1=O