CAS RN 99032-71-0
CAS RN 99032-71-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
1-Linoleoyl-3-palmitoyl-rac-glycerol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.