CAS RN 99199-62-9
CAS RN 99199-62-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
rac 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
rac 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
rac 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-methanol, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.