CAS RN 99347-76-9
CAS RN 99347-76-9
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
2-[[1-[(1,4-Dimethoxy-3-methyl-2-naphthalenyl)methyl]-2-methyl-2-propen-1-yl]thio]-acetic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[[1-[(1,4-Dimethoxy-3-methyl-2-naphthalenyl)methyl]-2-methyl-2-propen-1-yl]thio]-acetic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2-[[1-[(1,4-Dimethoxy-3-methyl-2-naphthalenyl)methyl]-2-methyl-2-propen-1-yl]thio]-acetic Acid Methyl Ester, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.