CAS RN 99533-80-9
CAS RN 99533-80-9
IUPAC Nome:
methyl (15S,18R)-16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Sinonimi:
methyl (15S,18R)-16-hydroxy-15-methyl-3-oxo-28-oxa-4,14,19-triazaoctacyclo[12.11.2.1¹⁵,¹⁸.0²,⁶.0⁷,²⁷.0⁸,¹³.0¹⁹,²⁶.0²⁰,²⁵]octacosa-1(26),2(6),7(27),8(13),9,11,20,22,24-nonaene-16-carboxylate
Peso molecolare (g/mol):
467.48
Formula molecolare:
C27H21N3O5
InChi Key:
KOZFSFOOLUUIGY-UHOLRESSSA-N
SMILES:
COC(=O)C1(O)C[C@H]2O[C@]1(C)N1C3=C(C=CC=C3)C3=C1C1=C(C4=CC=CC=C4N21)C1=C3CNC1=O
1
–
4
di
4
risultati
1
K252a, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
K252a, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
K252a, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.