CAS RN 99584-96-0
CAS RN 99584-96-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
5
di
5
risultati
1
DBHDA, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
DBHDA, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Promozioni disponibili
Tocris Bioscience™ DBHDA
Tocris Bioscience™ DBHDA
Reagent for synthetic biology; converts cysteine to dehydroalanine
4
DBHDA, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5