CAS RN 996-12-3
CAS RN 996-12-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
2,2-dimethylhexanal, TRC
CAS: 996-12-3 Formula molecolare: C8H16O Molecular Weight (g/mol): 128.21 IUPAC Name: 2,2-dimethylhexanal SMILES: CCCCC(C)(C)C=O