UserName
Lab Continuity And Preparedness Lab Equipment
How to Choose the Best Nitrile Isolator/RABS Gloves for Your Needs
Marzo 2023 | 2 min read | By: Ansell
How to Choose the Best Nitrile Isolator/RABS Gloves for Your Needs
For over 100 years, Ansell™ has been ensuring workplace safety with a broad range of innovative solutions, protecting countless workers around the globe. When it comes to choosing the best gloves to suit your needs, it is important to select a partner you can trust and consider all the products and solutions on offer. 
"However, which gloves should you use for your specific application? The answer to this question hinges on a number of factors, not least the type of hazard faced and the level of protection needed."
Which Nitrile Isolator/RABS Gloves Should I Choose?
Isolator/RABS gloves and their glove boxes play a vital role in protecting wearers and their products from contamination in cleanroom environments, safeguarding both employee well-being and product integrity in the face of potential workplace hazards. Our high-quality BioClean™ Nitrile Isolator/RABS portfolio consists not only of gloves, but also mittens, sleeves, and sleeve/glove systems, all of which are individually triple-bagged in easy-to-open polyethylene (PE) to enable simple, convenient access. They are truly the best choice for workers seeking the perfect blend of comfort, protection, and performance. 
BioClean Nitrile Isolator/RABS Gloves: What Sets Them Apart?
Nitrile is a synthetic, non-solvent based, FDA-approved polymer and is an ideal material for parenteral drug manufacturing. With excellent anti-static properties preventing the buildup of static electricity, nitrile is ideal for use with solvents and powders. It can be sterilized by gamma irradiation and sanitized with vaporized hydrogen peroxide (VHP) and isopropyl alcohol (IPA) and is autoclave compatible.
BioClean™ Nitrile Isolator/RABS gloves are washed, processed, and packaged within a cleanroom environment, ensuring the gloves are an ultra-low contamination risk before being introduced into the isolator glove box. Resistant to chemical hazards of varying severity (including cytotoxins), BioClean™ gloves are also powder-free and latex-free, resulting in greater resilience and a lesser risk of Type IV latex allergies. Their optimal comfort levels are coupled to a robust nitrile formulation that can withstand punctures and tears, making them the obvious choice for workers who require reassurance and genuine peace of mind.
If you’re looking for isolator/RABS gloves that offer the highest level of protection, look no further than the Ansell™ BioClean™ Nitrile range.
In our new video we show you why, when choosing our high-quality nitrile isolator/RABS gloves, you are choosing the best protective gloves out there, not only for yourself but also your products.