UserName
Sustainability Lab Equipment
Which Technologies Make it Possible to Reduce Energy Consumption and Environmental Impact with High Performance Floor Model Centrifuges?
Settembre 2023 | 2 min read | By: Thermo Scientific
Which Technologies Make it Possible to Reduce Energy Consumption and Environmental Impact with High Performance Floor Model Centrifuges?
Introduction
Superspeed centrifuges are critical instruments in both basic research and bioprocessing applications, and their continuous use can significantly contribute to the overall energy consumption of a lab. To manage energy consumption and environmental impact in high-use environments, the Thermo Scientific™ Sorvall™ LYNX 4000 and LYNX 6000 superspeed centrifuges feature a Green Mode, enabling the centrifuge to sleep when not in use for more than 2 hours. With the Sorvall LYNX 6000 centrifuge, further energy savings are made possible with its Smart Vacuum feature. Additionally, these centrifuges comply with Restriction of Hazardous Substances (RoHS) and Waste Electrical and Electronic Equipment (WEEE) directives, and are packaged in environmentally-friendly materials.
Green Mode
When not in use for more than 2 hours, a Sorvall LYNX superspeed centrifuge can be set to sleep via its Green Mode. A simple touch on the control panel wakes the centrifuge for immediate use. During low-use periods, such as nights and weekends, this can save significant power – up to 64% when compared to our previous generation of Thermo Scientific superspeed centrifuges (figure 1).
24690-table1.jpg
Figure 1. Comparison of Energy Use with and without Green Mode (for an Idle Centrifuge)
Smart Vacuum
Smart Vacuum, available on the Sorvall LYNX 6000 centrifuge, removes up to 80% of the air inside the centrifuge chamber. As a result, air friction on the spinning rotor is greatly reduced, and the power required to spin and cool the rotor is decreased by up to 74% when compared to identical run conditions on our previous generation of Thermo Scientific superspeed centrifuges (figure 2).
24690-table2.jpg
Figure 2. Comparison of Energy Use with and without Smart Vacuum (for 6 x 1000mL Rotors Run at 8,500rpm and 4°C)
Eco-Friendly Product Packaging
Superspeed centrifuges are packaged in recycled corrugated cartons with a biodegradable protective cover. Additionally, the fresh wood fiber used for transportation is produced on sustainable plantations where each tree cut down is replaced by two new plants, supporting forest ecosystem management.
Summary
In both high-use research and bioprocessing environments, Thermo Scientific Sorvall LYNX Superspeed Centrifuges provide substantial energy savings due to their Green Mode and Smart Vacuum options, as well as enhanced sustainability due to their eco-friendly packaging. Discover more about how you can simplify performance, save energy, and improve sustainability.