UserName
New Innovative Technology Lab Equipment
HABITAT Ferment: The New Smart Laboratory Bioreactor from IKA
Novembre 2025 | 2 min read | By: IKA
HABITAT Ferment: The New Smart Laboratory Bioreactor from IKA
A New Standard in Scalable Bioprocessing
With over a century of expertise in laboratory technology, IKA™ brings you the HABITAT Research Bioreactor, a state-of-the-art platform that merges tradition with innovation. Engineered for maximum performance, HABITAT sets new benchmarks in mixing efficiency, temperature control, automation, user safety, and ergonomic design.
Designed for maximum flexibility, HABITAT is ideal for cell culture, microbial fermentation, or phototrophic cultivation. Its compact benchtop footprint makes it a perfect fit for research laboratories seeking high-performance capabilities in a space-saving format.
Three Functions, One System, Unlimited Potential
Whether you're cultivating mammalian cells, microbial cultures, or algae, HABITAT seamlessly transforms into a bioreactor, fermenter, or photobioreactor. Pair it with IKA thermostats or optional LED-RGB light panels to suit your specific process needs, all from a single, trusted provider.
Available with single- or double-walled vessels in volumes from 0.5 to 10 L, the HABITAT Research Bioreactor offers a scalable solution tailored to your workflow, from small-scale experimentation to process optimization.
25677_IKA_article.jpg
Your Advantages with the IKA HABITAT
One System with Endless Bioprocessing Possibilities
  • Versatile bioreactor platform: Designed for optimal growth of any cell type, from microbial to mammalian and plant cells
  • Integrated gas management: Each tower package includes a configurable mass flow controller for air, N₂, O₂, and CO₂ ensuring precise gas delivery
  • Built-in fluid handling: Every tower package comes equipped with four bidirectional peristaltic pumps for flexible media and reagent management
Advanced Sensor Integration
  • Comprehensive sensor suite: pH, DO, temperature, anti-foam, and level sensors are standard in every vessel package
  • Upgrade with high-tech options: Integrate cutting-edge sensors such as viable biomass sensors, optical DO, redox, or off-gas analyzers
  • Data-driven process insight: Fully compatible with soft sensors for advanced data analysis and process optimization
Intuitive and Powerful Control Software
  • Effortless operation: User-friendly interface simplifies setup and day-to-day operation.
  • Automated process control: Fully automated regulation of critical parameters like pH and temperature.
Exclusive Features that Set Us Apart
  • 3-in-1 flexibility: Use as a Bioreactor, Fermenter, or Photobioreactor – all in a single platform
  • Virtual reality training: Train personnel in a safe, immersive digital environment – perfect for onboarding and education
  • Innovative lid stand: Enhances workflow by eliminating the need to remove and handle the lid during setup
  • Chaotic mixing mode: A novel, chaos-based mixing algorithm ensures rapid, homogenous blending beyond traditional methods