Heating and Drying Ovens
Heating and drying ovens are used for simultaneously heating and drying samples. Features include gravity or mechanical (forced air) convection, capacity, achievable temperature, programmability, and scheduled on/off cycles. Applications include drying, baking, aging tests, glassware drying, dry sterilization, and processing electronics.
Thermo Scientific™ Heratherm™ General Protocol Ovens - 230VAC 50/60Hz
Carry out daily laboratory work with Thermo Scientific™ Heratherm™ General Protocol Ovens, featuring temperature uniformity, easy settings and low energy consumption.
Contec™ Sterile IPA
BINDER™ Series FD-S Solid.Line - Drying and Heating Chamber, with Forced Convection
The Solid.Line is suited to drying and heating. The attributes of the Solid.Line are geared towards the application in question and therefore used in research and quality assurance.
Contec™ Sterile IPA
IKA™ Oven 125 Basic Dry Glass with UK Plug
Universal oven for tempering, drying, aging and heating application in the laboratory.
Contec™ Sterile IPA
Complete Your Order - Great Deals
Standard Incubators
Standard incubators provide precise temperature control and uniformity for a range of routine applications. Choose from a variety of sizes, airflow technologies, and fan speeds, as well as assorted program and display options, alarms, and general fitting variations according to your needs.
Lorem Ipsum
Dry Block Incubators
Dry block incubators are used to control temperature stability and uniformity in a range of applications. Modular systems provide versatility while digital models offer touchscreen options. Various form factors, temperature ranges, and shaking or mixing capabilities are available to meet your needs.
Lorem Ipsum
CO2 Incubators
Discover precise CO2 incubators for cell growth, tissue culture, stem cell research, and IVF. Our incubators ensure accurate conditions with advanced sensors and user-friendly interfaces. Choose our incubators for efficiency, safety, and consistency. Shop now to enhance your research!
Lorem Ipsum
FAQ
A laboratory heating or drying oven is designed to provide controlled, uniform temperatures for applications such as drying glassware, removing moisture from samples, sterilizing instruments, and conducting thermal testing. They are widely used in research, quality control, and industrial laboratories.
Natural convection ovens rely on rising hot air to circulate heat, which is ideal for gentle drying and reduced sample disturbance. Forced convection ovens use a built-in fan to provide faster, more uniform heating, making them better suited for higher throughput or when consistent drying conditions are critical.
Important factors include temperature range and stability, chamber size, safety features (such as over-temperature protection), air circulation (natural vs. forced convection), and whether vacuum or inert gas operation is needed. Accessories like additional shelves, data logging, or inert gas kits can also improve usability.
Resources and Related Information
Control the elements - Thermo Scientific heating equipment
Thermo ScientificTM products deliver reliable temperature, humidity, and light conditions to meet the needs of your application.
Lorem Ipsum
Top Brands