Gloves That Set the Benchmark for Safety and Integrity
An acceptable quality level (AQL) both establishes testing criteria and assesses product quality in manufacturing. The lower the number, the lower incidence of defects and thus the higher the quality of the item produced. In critical environments, such as in industries like pharmaceuticals, biotech and semiconductors, an AQL of 1.5 is typically the norm.
Biogel™ Tech Gloves offer unmatched quality and, with an industry-leading AQL of 0.4, all our gloves exceed the highest standards to deliver superior protection for both operators and products. Trust the gloves that set the benchmark for safety and integrity and experience the unmatched quality of Mölnlycke™.
Content provided by: