Concentration of solutions
Percent Composition (by mass)
You can consider percent by mass (or weight percent) in two ways:
- The parts of solute per 100 parts of solution
- The fraction of a solute in a solution multiplied by 100
You need two pieces of information to calculate the percent by mass of a solute in a solution:
- The mass of the solute in the solution
- The mass of the solution
Use the following equation to calculate percent by mass:
Mass Percentage = (Solute mass/Solution mass) X 100
Molarity
Molarity tells us the number of moles of solute in exactly one litre of a solution. (Note that molarity is represented by a capital M).
You need two pieces of information to calculate the molarity of a solute in a solution:
- The moles of solute present in the solution
- The volume of solution (in litres) containing the solute
To calculate molarity use the equation:
Molarity = Moles of solute/Volume of solution (L)
Molality
Molality, m, tells us the number of moles of solute dissolved in exactly one kilogram of solvent. (Note that molality is represented by a lower case m).
You need two pieces of information to calculate the molality of a solute in a solution:
- The moles of solute present in the solution
- The mass of solvent (in kilograms) in the solution
To calculate molality we use the equation:
Molality = Moles of solute/Mass of solvent (Kg)
Normal and Molar Soluions
| Concentrated reagents | Specific gravity | Molarity | Normality | Volume (mL) required to make 1L of solution | |
| SG | M | N | 1M | 1N | |
| Acetic acid 99,5% | 17,40 | 17,40 | 57,5 | 57,5 | |
| Ammonia 35% | 0,88 | 18,10 | 18,10 | 55,3 | 55,3 |
| Ammonia 25% | 0,91 | 13,40 | 13,40 | 74,6 | 74,6 |
| Hydrochloric acid 36% | 1,18 | 11,65 | 11,65 | 85,8 | 85,8 |
| Hydrochloric acid 32% | 1,16 | 10,20 | 10,20 | 98,0 | 98,0 |
| Hydrofluoric acid 40% | 1,13 | 22,60 | 22,60 | 44,2 | 44,2 |
| Nitric acid 70% | 1,42 | 15,80 | 15,80 | 63,3 | 63,3 |
| Perchloric acid 60% | 1,54 | 9,20 | 9,20 | 108,7 | 108,7 |
| Perchloric acid 70% | 1,67 | 11,60 | 11,60 | 86,2 | 86,2 |
| Orthophosphoric acid 85% | 1,70 | 15,20 | 45,60 | 65,8 | 21,9 |
| Sodium hydroxide 47% | 1,50 | 17,60 | 17,60 | 56,7 | 56,7 |
| Sulfuric acid 96% | 1,83 | 18,00 | 36,00 | 53,1 | 26,2 |