CAS RN 943430-62-4
CAS RN 943430-62-4
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
4,7-Dichloro-2-phenyl-1H-isoindole-1,3(2H)-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4,7-Dichloro-2-phenyl-1H-isoindole-1,3(2H)-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.