CAS RN 946821-59-6
CAS RN 946821-59-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
10-O-Acetyl SN-38, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
10-O-Acetyl SN-38, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
10-O-Acetyl SN-38, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.