CAS RN 948304-40-3
CAS RN 948304-40-3
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Promozioni disponibili
Tocris Bioscience™ t-TUCB
Tocris Bioscience™ t-TUCB
Potent soluble epoxide hydrolase (sEH) Inhibitor; anti-inflammatory and antinociceptive