CAS RN 952309-99-8
CAS RN 952309-99-8
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
N-des-(3-cyclohexen-1-ylmethyl)-N-ethylcarbamoyl Cinitapride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-des-(3-cyclohexen-1-ylmethyl)-N-ethylcarbamoyl Cinitapride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.