CAS RN 952686-58-7
CAS RN 952686-58-7
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
2-[[(5-Nitro-2-thiazolyl)amino]carbonyl]phenyl 2-(acetyloxy)benzoate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.