CAS RN 956903-21-2
CAS RN 956903-21-2
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
(2R,3R,11bR)-rel-9-deMe-DTBZ, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(2R,3R,11bR)-rel-9-deMe-DTBZ, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.