CAS RN 95748-23-5
CAS RN 95748-23-5
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
2-(3,4-Dimethoxyphenyl)-5-{[2-(3,4-dimethoxyphenyl)-ethyl]methyl-amino}-pentanenitrile, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.