CAS RN 96115-88-7
CAS RN 96115-88-7
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
1
di
1
risultati
1
Carbidopa Ethyl Ester Hydrochloride Salt, TRC
CAS: 96115-88-7 Formula molecolare: C12H19ClN2O4 Molecular Weight (g/mol): 290.74 Sinonimo: alpha-Hydrazinyl-3,4-dihydroxy-alpha-methylbenzenepropanoic Acid Ethyl Ester Hydrochloride,alpha-Hydrazino-3,4-dihydroxy-alpha-methylhydrocinnamic Acid Ethyl Ester Hydrochloride IUPAC Name: ethyl 3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoate SMILES: Cl.CCOC(=O)C(C)(Cc1ccc(O)c(O)c1)NN