CAS RN 97760-87-7
CAS RN 97760-87-7
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
1-[4-Amino-3-chloro-5-(trifluoromethyl)phenyl]-2-bromo-ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1-[4-Amino-3-chloro-5-(trifluoromethyl)phenyl]-2-bromo-ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1-[4-Amino-3-chloro-5-(trifluoromethyl)phenyl]-2-bromo-ethanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.