CAS RN 98474-59-0
CAS RN 98474-59-0
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
3
di
3
risultati
1
MedChemExpress Myelin Basic Protein (MBP) (68-82), guinea pig
MedChemExpress Myelin Basic Protein (MBP) (68-82), guinea pig is a fragment of myelin basic protein (MBP).
2
MedChemExpress Myelin Basic Protein (MBP) (68-82), guinea pig
MedChemExpress Myelin Basic Protein (MBP) (68-82), guinea pig is a fragment of myelin basic protein (MBP).
3
MedChemExpress Myelin Basic Protein (MBP) (68-82), guinea pig
MedChemExpress Myelin Basic Protein (MBP) (68-82), guinea pig is a fragment of myelin basic protein (MBP).