CAS RN 98818-49-6
CAS RN 98818-49-6
IUPAC Nome:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
(AlphaR,DeltaS,GammaR)-Delta-[[(1,1-Dimethylethoxy)carbonyl]amino]-Gamma-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-Alpha-(phenylmethyl)benzenehexanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(AlphaR,DeltaS,GammaR)-Delta-[[(1,1-Dimethylethoxy)carbonyl]amino]-Gamma-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-Alpha-(phenylmethyl)benzenehexanoic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.