Standard Incubators for Your Laboratory
Selecting the right standard incubator is essential for ensuring reliable and reproducible results in microbiology, pharmaceutical, and research laboratories. With multiple configurations, temperature ranges, and airflow systems available, choosing the right model depends on your specific application requirements.
Useful Links
Save Now - Exclusive Deals
Product Code 10529070
Product Code 12815883
Product Code 15682056
Product Code 10839810
Product Code 15662056
Product Code 15810312
Product Code 15652056
Complete Your Order - Great Deals
Must Have
Product Code 15672116
Product Code 15612126
Product Code 12625306
Product Code 15622116
FAQ
A standard incubator is a laboratory device designed to maintain a controlled temperature environment for the incubation of samples. It is commonly used for microbiological cultures, sample storage, and stability testing.
Unlike CO₂ incubators, standard incubators do not control gas levels, making them suitable for general-purpose applications where precise atmospheric conditions are not required.
Temperature Range and Stability
The required temperature range depends on your application. Most standard incubators operate between ambient +5°C and 70°C. Stability and uniformity are critical to ensure consistent experimental conditions.
Chamber Volume and Capacity
Choose a chamber size based on your sample throughput. Larger capacities are ideal for high-volume labs, while compact units suit limited bench space.
Air Circulation Technology
- Natural convection: gentle airflow, reduces sample drying
- Forced air convection: faster temperature recovery and better uniformity
Control and Monitoring
Digital controls, alarms, and data logging features improve reliability and compliance with laboratory standards.
| Type | Key Feature | Typical Use |
|---|---|---|
| Standard incubator | Temperature control only | Microbiology, general lab use |
| CO₂ incubator | Gas + humidity control | Cell culture |
| Refrigerated incubator | Cooling + heating | BOD testing, environmental studies |
Understanding these differences helps ensure you select the right equipment for your application.
Standard incubators are widely used for:
- Microbiological culture growth
- Pharmaceutical stability testing
- Food and beverage quality control
- Research and academic laboratories