Learn More
1,1,1,3,3,3-Hexafluoro-2-methoxypropane, Mikromol™
Product Code. 17904316
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17904316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17904316 Supplier Mikromol Supplier No. MM3315.020100
 Restricted Item
Spedizione stimata: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1,1,1,3,3,3-Hexafluoro-2-methoxypropane
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1,1,3,3,3-hexafluoro-2-methoxy-propane
CAS 13171-18-1
Analyte or Component Names 1,1,1,3,3,3-hexafluoro-2-methoxy-propane
Shipping Condition Room Temperature
Molecular Formula C4 H4 F6 O
Certifications/Compliance ISO 9001
Synonym 0Sevoflurane Related Compound B, Sevoflurane Impurity B
InChI Formula InChI=1S/C4H4F6O/c1-11-2(3(5,6)7)4(8,9)10/h2H,1H3
SMILES COC(C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,3,3,3-hexafluoro-2-methoxypropane
Molecular Weight (g/mol) 182.06
Quantity 100 mg
Formula Weight 182.0166 g/mol