missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Diphenylethylene, 98%, Thermo Scientific Chemicals
Click to view available options
Quantità:
10 g
25 g
100 g
250 g
Descrizione
1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.
Solubility
Miscible with methanol, chloroform and ether. Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents
1,1-Diphenylethylene is used in the preparation of 2-chloro-1,1-diphenyl-ethene by reacting with benzeneseleninyl chloride and aluminum(III) chloride as reagents. It acts as an intermediate in organic synthesis as well as in pharmaceuticals.
Solubility
Miscible with methanol, chloroform and ether. Insoluble in water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents
Specifica
Materiale o nome chimico | 1,1-Diphenylethylene |
Punto di fusione | 5°C to 7°C |
Densità | 1.023 |
Punti di ebollizione | 270°C to 271°C |
Punto d'infiammabilità | 136°C (276°F) |
Indice di rifrazione | 1.6065 |
Quantità | 25 g |
Beilstein | 1099062 |
Indice di Merck | 14,3323 |
Informazioni di solubilità | Miscible with methanol,chloroform and ether. Insoluble in water. |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'