missing translation for 'onlineSavingsMsg'
Learn More

1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals

Catalog Number p-3737172
Click to view available options
Quantità:
5 g
25 g
100 g
500 g
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5144-89-8, 66-71-7
C12H10N2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
21226
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5144-89-8, 66-71-7
C12H10N2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
21226
O.C1=CN=C2C(C=CC3=CC=CN=C23)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 1, 10-Phenanthroline monohydrate
CAS min. % 99.0
CAS max % 100.0
Punto di fusione 97.0°C to 101.0°C
Colore Cream-White
Spettro a infrarossi Authentic
Intervallo percentuale saggio 99+%
Imballaggio Glass bottle
Quantità 5 g
Beilstein 23,227
Indice di Merck 15,7328
Sinonimo 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate
Informazioni di solubilità Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone and benzene
IUPAC Name 1,10-phenanthroline;hydrate
Peso formulazione 198.23
Percent Purity 99+%
Grado Pure
Forma fisica Needle-like Crystalline Powder
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'