missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Cyclobutanetetracarboxylic Acid 1,2,3,4-Tetramethyl Ester, TRC

Codice prodotto. 30010244
Cambia vista
Click to view available options
Quantità:
50 mg
100 mg
250 mg
Dimensione della confezione:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
30010244 100 mg 100mg
30134573 250 mg 250mg
30010039 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30010244 Fornitore Toronto Research Chemicals N. del fornitore TRCC989408100MG
 Articolo soggetto a restrizioni
Spedizione stimata: 11-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 14495-41-1
Formula molecolare C12 H16 O8
Molecular Weight (g/mol) 288.25
Sinonimo 1,2,3,4-Cyclobutanetetracarboxylic acid, 1,2,3,4-tetramethyl ester, 1,2,3,4-Cyclobutanetetracarboxylic acid, tetramethyl ester (6CI,7CI,8CI,9CI), Tetramethyl cyclobutane-1,2,3,4-tetracarboxylate
IUPAC Name tetramethyl cyclobutane-1,2,3,4-tetracarboxylate
SMILES COC(=O)C1C(C(C1C(=O)OC)C(=O)OC)C(=O)OC

Specifica

Materiale o nome chimico 1,2,3,4-Cyclobutanetetracarboxylic Acid 1,2,3,4-Tetramethyl Ester
Note sul grado di purezza HPLC
Quantità 100 mg
InChI Formula InChI=1S/C12H16O8/c1-17-9(13)5-6(10(14)18-2)8(12(16)20-4)7(5)11(15)19-3/h5-8H,1-4H3
Peso formulazione 288.0845
Percent Purity >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.