Learn More
1-(2-Chlorophenyl)piperazine Hydrochloride, TRC
Product Code. 30362243
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362243 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30362243 Supplier Toronto Research Chemicals Supplier No. TRCC350685500MG
 Restricted Item
Spedizione stimata: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material ortho-Chlorophenylpiperazine Hydrochloride
CAS 41202-32-8
Molecular Formula C10 H13 Cl N2 . Cl H
Quantity 500 mg
Synonym ortho-Chlorophenylpiperazine Hydrochloride, oCPP HCl
InChI Formula InChI=1S/C10H13ClN2.ClH/c11-9-3-1-2-4-10(9)13-7-5-12-6-8-13;/h1-4,12H,5-8H2;1H
SMILES Cl.Clc1ccccc1N2CCNCC2
IUPAC Name 1-(2-chlorophenyl)piperazine;hydrochloride
Molecular Weight (g/mol) 233.14
Formula Weight 232.0534