Learn More
1,2-Dimethylimidazole, 98%, Thermo Scientific Chemicals
Catalog Number p-7027076
13.53 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.
Click to view available options
Quantità:
25 g
100 g
500 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1739-84-0
C5H8N2
96.13
MFCD00005294
GIWQSPITLQVMSG-UHFFFAOYSA-N
1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole
15617
1,2-dimethylimidazole
CN1C=CN=C1C
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1739-84-0
C5H8N2
96.13
MFCD00005294
GIWQSPITLQVMSG-UHFFFAOYSA-N
1,2-dimethyl-1h-imidazole, 1h-imidazole, 1,2-dimethyl, imidazole, 1,2-dimethyl, 1h-imidazole, dimethyl, unii-q41bc3grjb, q41bc3grjb, lupragen r dmi, 1,2-dimethylimdazole, 1,2 dimethylimidazole, n,2-dimethylimidazole
15617
1,2-dimethylimidazole
CN1C=CN=C1C
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Dimethylimidazole is used in the synthesis of 1,2-dimethyl-3-n-butylimidazoliumchloride and 1,2-dimethyl-3-n-propylimidazolium chloride. It also can be used in the synthesis of 1-(2-methoxyethyl)-2,3-dimethylimidazolium chloride and hexafluorophosphate salts.
Solubility
Soluble in water.
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico 1,2-Dimethylimidazole
Punto di fusione 35°C to 38°C
Densità 1.084
Punti di ebollizione 203°C to 205°C
Punto d'infiammabilità 92°C (197°F)
Quantità 25 g
Beilstein 108457
Informazioni di solubilità Soluble in water.
Peso formulazione 96.13
Percent Purity 98%
compliance-icons compliance-icons
Avvertenza
  • Pericolo
Categoria di pericolo
  • Tossicità acuta Categoria 4
  • Lesione oculare grave/irritazione oculare Categoria 1
  • Corrosione/irritazione della pelle Categoria 2
  • Sensibilizzazione della pelle Categoria 1
Indicazioni di pericolo
  • H302-Nocivo se ingerito.
  • H315-Provoca irritazione cutanea.
  • H317-Può provocare una reazione allergica cutanea.
  • H318-Provoca gravi lesioni oculari.
Consigli di prudenza
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P301+P330+P331-IN CASO DI INGESTIONE: sciacquare la bocca. NON provocare il vomito.
  • P302+P352-IN CASO DI CONTATTO CON LA PELLE: lavare abbondantemente con acqua/.