Learn More
1-[(2S)-2-Amino-1-oxo-3-phenylpropyl]pyrrolidine, TRC
Codice prodotto. 30325237
Cambia vista
Click to view available options
Quantità:
250 mg
Dimensione della confezione:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30325237 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30325237 Fornitore Toronto Research Chemicals N. del fornitore TRCA618760250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico 1-[(2S)-2-Amino-1-oxo-3-phenylpropyl]pyrrolidine
CAS 56414-89-2
Formula molecolare C13 H18 N2 O
Quantità 250 mg
InChI Formula InChI=1S/C13H18N2O/c14-12(10-11-6-2-1-3-7-11)13(16)15-8-4-5-9-15/h1-3,6-7,12H,4-5,8-10,14H2/t12-/m0/s1
SMILES N[C@@H](Cc1ccccc1)C(=O)N2CCCC2
IUPAC Name (2S)-2-amino-3-phenyl-1-pyrrolidin-1-ylpropan-1-one
Molecular Weight (g/mol) 218.29
Peso formulazione 218.1419